Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Steri-Wrap LS Series Paper, CRP, KNF

Supplier: KNF

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

600061-02 600061-03
12243-852EA 6774.33 USD InStock
12243-852 12243-850
Steri-Wrap LS Series Paper, CRP, KNF
Sterilization Wraps
Nomex nylon paper is made from polyamide polymers and offers excellent mechanical, thermal and nonlinting properties.

  • Withstands temperatures in excess of 260°C (500°F)
  • Core size of 7.6 cm (3").
  • 2 mil. paper thickness

Ideal sterile wrap for pharmaceutical and biological products, and medical devices.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty