Would you like to log in again?
Hanging Pan Balances, Ohaus
Supplier: Ohaus
80000009 80000010
12338-026EA 553.47 USD InStock
12338-026 12337-001
Hanging Pan Balances, Ohaus
Balances and Scales
Perfect for general laboratory weighing
Stainless steel pan and bow, tiered and notched aluminum beams, precision ground knives, covered agate bearings, and parallax-free end reading. Designed to read accurately even when out of level. Balances feature magnetic damping and self-storing specific gravity platform. The Dial-O-Gram® has an engraved dial and vernier; the Cent-O-Gram® provides four-beam weight adjustment.
Ordering information: Supplied with manufacturer’s five-year warranty.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty