Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Bel-Art Petri Dish Plate Reader, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F37861-0000
12574-064EA 312.67 USD InStock
12574-064
SP Bel-Art Petri Dish Plate Reader, Bel-Art Products, a part of SP
Microplate Readers
This reader allows hands-free viewing of Petri dishes through a large, adjustable mirror with 3× magnification.
  • Polypropylene reader construction with an acrylic deck
  • Safe examination of dishes
Ideal for hemagglutination, hemagglutination inhibition, hemolysis, and agglutination testing. Other applications include counting of cell cultures for colony or plaque growth. The reader assists in evaluation of monolayer cell growth in Petri dishes, viewing of color changes, and locating cells in microtiter plates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty