Would you like to log in again?
VWR® Weighing Paper
Supplier: VWR
VWR®
20 60 5628 20 60 5626 20 60 5627
12578-201CS 2318.01 USD InStock
12578-201 12578-121 12578-165
VWR® Weighing Paper
Weighing Papers
Weighing paper is designed for analytical weighing with minimal influence on analytical results for weighing and transporting sample.
  • Clean and dust-free
  • Lightweight
  • Nitrogen-free
  • Smooth surface
Smooth surface is ideal for transferring weighed material. Clean, dust-free sheets. Paper is lightweight, yet strong enough to support analytical size samples.
Packaging: All sizes of weighing papers are packaged 500 sheets per box.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty