Would you like to log in again?
VWR® Signature™ Incubating and Cooling Orbital Shaker
Supplier: VWR
VWR® Signature™
80863459 80863458
12620-934EA 7711 USD InStock
12620-934 10027-136
VWR® Signature™ Incubating and Cooling Orbital Shaker
Shakers and Mixers Shaking Incubators
Ideal for cell cultures, hybridizations, and suspensions.
  • Optimized for shaking microplates, tubes, and vials
  • Digital display for speed, temperature, and time
  • Microprocessor control
  • Timer with audible alarm
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Easy-to-use controls allow user to adjust temperatures in 1 °C increments
Microprocessor PID temperature controller maintains precise temperature control from 10° below ambient to 65 °C (149 °F). The triple eccentric drive features permanently lubricated ball bearings. The maintenance-free, brushless DC motor provides a long service life. A Lexan® polycarbonate lid permits viewing of samples from all sides without disturbing internal temperature.
Shaker holds two microplates or two modular blocks (sold separately). See website for modular blocks capable of accommodating up to 18 different tube or vial configurations.
Built-in timer with LED display will show elapsed time, or, when programmed to user-defined limit, will count down to zero and shut off unit. Audible alarm sounds when time reaches zero.
Certifications: RoHS certified. CSA listed. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the temperature and speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Unit includes a 2.3m (92") detachable, three-wire cord and plug. 230V, 50/60Hz models are also available; contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty