Would you like to log in again?
VWR® Incubating Mini Shaker
Supplier: VWR
VWR®
80863463 80863462
10027-124EA 6661.4 USD InStock
10027-124 12620-942
VWR® Incubating Mini Shaker
Shakers and Mixers Shaking Incubators
Variable-speed, microprocessor control provides consistent, uniform mixing action.
  • Digital Display for Speed, Temperature, and Time
  • Microprocessor Control
  • Timer with Audible Alarm
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Ramping feature slowly increases speed to set point for added safety
The triple eccentric drive features permanently lubricated ball bearings. The maintenance-free, brushless DC motor provides a long service life. A Lexan® polycarbonate lid permits viewing of samples from all sides without disturbing internal temperature. Included tray can be used as a platform for mounting Erlenmeyer flask clamps, test tube racks, or microplate holders.
Independent LED display features speed, temperature, and time. Easy-to-use controls allow users to adjust temperature in 1 °C increments. Temperature calibration mode allows users to calibrate the unit to an external temperature device. Timer will display elapsed time, or, when programmed to user-defined limit, will count down to zero and shut off unit. Audible alarm sounds when time reaches zero. Durable base is constructed of cast aluminum for extra stability.
Certifications: RoHS certified. CSA Listed. Calibrated Shakers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the temperature and speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Units include a 2.3m (7.5') detachable, 3-wire cord and plug. 230V, 50/60Hz units are also available; contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty