Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Multi-Position Magnetic Stirrers, 120 V
Supplier: VWR
VWR®
80864013 80864011 80864003 80864002 80864006 80864010 80864014 80864009 80864007 80864001 80863999
10027-068EA 3065.87 USD InStock
10027-068 12621-018 10027-078 12621-038 10027-076 12621-026 12621-034 12621-046 10027-070 12621-022 12621-042
VWR® Multi-Position Magnetic Stirrers, 120 V
Stirrers Magnetic Stirrers, Multi-position
Powerful magnetic stirrers with precise speed control offer repeatable performance.
  • Stir up to 9 vessels at once
  • Microprocessor speed control
  • Synchronized, uniform mixing
  • Low-profile design
  • Now with Best-in-Class, Exclusive 5-Year Warranty
Synchronized operation ensures uniform mixing at each position
Model 650 standard series offers an analog speed control knob with dial markings. Model 750 advanced series offers an LED display for digital speed control, adjustable in 10rpm increments.
Ramping feature slowly brings sample to set speed to avoid splashing, improve magnetic coupling, and provide excellent low-speed stirring. Large top plate with position indicators is easy to clean and provides a white reflective surface. Low-profile design takes up less space and easily fits into fume hoods. Spill-resistant housing channels fluids away from internal components. Cast metal base offers durability and added stability.
Certifications: RoHS certified. UL and C-UL listed. Calibrated Stirrers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Units include a 2.3m (92") detachable three-wire electrical cord with plug. Units are also supplied with a 2.5 cm (1") PTFE-coated stir bar. 230V, 50/60Hz models are also available; contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty