Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Advanced and Standard Multi-Position Stirrers, 230 V (Export Only)

Supplier: VWR
VWR®
80864008 80864000 80864012 80864004 80864015
12621-028EA 2030.1 USD InStock
12621-028 12621-036 12621-048 12621-044 12621-020
VWR® Advanced and Standard Multi-Position Stirrers, 230 V (Export Only)
Stirrers Magnetic Stirrers, Multi-position
Microprocessor controlled
Synchronized and uniform mixing
Low-profile design
Best-in-Class, 5-year warranty
Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner.

  • 230 V model
  • Precise speed control
  • Cast metal base offers durability and added stability
  • Excellent low-speed stirring

Standard series offers an analog speed control knob with dial markings

Powerful magnetic stirrers with precise speed control offer repeatable performance. Synchronized operation ensures uniform mixing at each position. Ramping feature slowly brings sample to set speed to avoid splashing, improve magnetic coupling, and provide excellent low-speed stirring. Large top plate with position indicators is easy to clean and provides a white reflective surface. Low-profile design takes up less space and easily fits into fume hoods. Spill-resistant housing channels fluids away from internal components.

Advanced series offers an LED display for digital speed control, adjustable in 10rpm increments.

Certifications: RoHS certified. Calibrated Stirrers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.

Ordering information: Units include a 234 cm (92") Euro type plug. Units are also supplied with a 2.5 cm (1") PTFE-coated stir bar. Stirrers with NIST traceable calibration certificate provided by an ISO/IEC 17025 accredited laboratory. Multiple data points within the speed range and time function are reported on the certificate with the associated uncertainties.

Caution: 230 V model.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty