Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Model 1000 & 2000 Advanced Large Capacity Stirrers, 230 V (Export Only)
Supplier: VWR
VWR®
80864028 80864022
12621-064EA 7289.07 USD InStock
12621-064 12621-060
VWR® Model 1000 & 2000 Advanced Large Capacity Stirrers, 230 V (Export Only)
Stirrers Magnetic Stirrers
Ideal for high volume applications, the powerful magnetic drive of this stirrer is capable of mixing high viscosity materials.
  • 230 V model
  • LED displays for speed and time
  • Durable stainless steel construction
  • Best-in-Class Five-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Stainless steel base offers durability and added stability.
Microprocessor control precisely regulates speed through the entire range. Stirring function, with continuous duty motor and magnet, maintains set speed even under changing load or viscosity. Speed ramping slowly increases speed for improved safety and enhanced coupling. Avoids splashing, improves spin bar control, and provides excellent low end speed control.
Certifications: RoHS Certified. TUV listed. CE Marked. Calibrated Stirrers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Ordering information: Unit includes a 234 cm (92") Euro type plug. Unit is also supplied with a 7 cm (2.75") egg-shaped PTFE-coated stir bar. Stirrers with NIST traceable calibration certificate provided by an ISO/IEC 17025 accredited laboratory. Multiple data points within the speed range and time function are reported on the certificate with the associated uncertainties.
Caution: 230 V model.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty