Would you like to log in again?
Individual Precision Analytical Weights, Class 3, Troemner
Supplier: Troemner
80858409 80858377 80858410 80858202 80858187 80858451 80858334 80858375 80858486 80858672 80858305 80858275 80858229 7016-3T 80858487 80858708 80858639 7013-3W 80858200 80858246 80858412 80858537 80858508 80858203 80858669 80858709 80858243 80858262 80858641 80858580 7013-3T 80858217 80858231 7010-3W 7014-3W 7012-3T 7010-3T 80858291 80858617 80858670 80858711 80858448 80858306 80858304 80858335 80858289 80858374 80858577 80858184 80858619 7014-3T 80858640 80858215 80858276 7014-3 80858507 80858277 80858449 80858245 80858337 80858232 80858509 7010-3 80858489 80858538 80858334 80858216 80858260 80858261 7012-3 80858578 80858540 80858616 80858290 80858186 7013-3 7016-3
14216-782EA 1045.22 USD InStock
14216-782 12765-390 12365-340 12765-350 12765-351 12765-352 12765-353 12765-354 12765-310 12765-355 12765-312 12765-356 12765-357 12765-314 12765-358 14216-780 12765-359 12765-349 12765-306 12765-308 12765-380 12765-381 12765-382 12765-383 14216-778 12765-384 12765-385 12765-386 12765-387 12765-300 12765-388 12765-389 12765-345 12765-346 12765-302 12765-347 14216-770 12765-304 12765-348 14216-802 14216-768 14216-804 14216-806 12765-370 12765-371 12765-372 12765-373 14216-766 12765-374 12765-375 12365-328 12765-376 12765-377 12365-326 12765-378 12765-379 12365-332 12365-330 14216-754 12765-360 14216-756 12765-361 12765-362 12765-363 12765-364 12765-320 12765-365 12365-338 12765-322 14216-790 12765-324 12365-336 14216-792 12365-334 12765-316 12765-318 AP400006-924
Individual Precision Analytical Weights, Class 3, Troemner
Weights Calibration Weights
Highly polished kilogram and gram weights made of Troemner Alloy 8, which is a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage
Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are aluminum); fractional weights 3 mg and below are made of aluminum.
This exclusive material has a density of 8.0 at 25 °C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to –195.5 °C (–320 °F).
Certifications: Standard weights supplied with a Statement of Accuracy. Where applicable, weights are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.
Order Now
ORDER
SERVICES
Services: VWR Catalyst™ Services provides convenient and fast re-calibration services within 6 business days for your weights. Please contact us at 1.888.793.2300 or VWRCATALYST@vwr.com for a quote or more information. If your weights are found to be out of tolerance, we will make the proper adjustments to bring them back into tolerance. To prevent loss and ensure the timely processing and integrity of your precision weights and/or calibration devices, please do not pack in foam peanuts. Please avoid packing your items with foam, packing peanuts or other material that cannot be recycled.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty