Would you like to log in again?
OIML Class F1 Precision Weights, Troemner
Supplier: Troemner
80780317 80780334 80780336 80780335 80780307 80780331 80780311 80780322 80780323 80780349 80780314 80780327 80780340 80780308 80780315 80780350 80780341 80780347 80780313 80780337 80780338 80780325 80780342 80780326 80780320 80780344 80780324 80780351 80780309 80780343 80780310 80780318 80780345 80780319 80780321 80780316 80780312 80780332 80780346 80780333 80780339 80780348
12766-016EA 303.51 USD InStock
12766-016 12766-038 12766-358 12766-336 12766-014 12766-036 12766-356 12766-034 12766-012 12766-354 12766-032 12766-010 12766-352 12766-030 12766-350 12766-370 12765-996 12766-348 12766-404 12766-008 12766-368 12766-346 12766-028 12766-402 12766-006 12766-344 12766-366 12766-004 12766-400 12766-364 12766-342 12766-002 12766-362 12766-340 12766-022 12766-000 12766-360 12766-020 12766-040 12765-998 12766-018 12766-338
OIML Class F1 Precision Weights, Troemner
Weights Calibration Weights
Troemner OIML Calibration Weights conform to the standards of the Organisation Internationale de Metrologie Legale International Recommendation (OIML R 111)
Weights 1 g and above are two-piece construction. Troemner OIML Calibration Weights 1 g and larger are cylindrical in shape with a tapered neck for easy gripping. They are perfectly flat on top to stack easily on center. Weight bottoms are slightly recessed to expose the smallest possible area to wear.
They are manufactured from the finest stainless steels available including Troemner Alloy 8 Stainless Steel that has low magnetic permeability, uniform consistency and high resistance to corrosion.
Order Now
ORDER
SERVICES
Services: VWR Catalyst™ Services provides convenient and fast re-calibration services within 6 business days for your weights. Please contact us at 1.888.793.2300 or VWRCATALYST@vwr.com for a quote or more information. If your weights are found to be out of tolerance, we will make the proper adjustments to bring them back into tolerance. To prevent loss and ensure the timely processing and integrity of your precision weights and/or calibration devices, please do not pack in foam peanuts. Please avoid packing your items with foam, packing peanuts or other material that cannot be recycled.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty