This easy-to-use, portable system integrates a variety of substance-specific chemical sensing chips with a handheld reader.
- Portable, easy to use
- Integrates variety of substance-specific, chemical-sensing chips with handheld reader
Flow sensor guarantees a constant mass flow of the sample through the capillary in the chip so that measurements are not affected by fluctuations in the atmospheric pressure. It takes gas and vapor spot measurements by combining chemistry and electronics to provide accurate and reliable results.
Ordering information: Chips are calibrated for up to two years. Analyzer can be equipped with a remote sampling unit. For details, contact your Avantor sales representative.