Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Sound Level Meter

Supplier: VWR
VWR®
93676-49
12777-836EA 601.68 USD InStock
12777-836
VWR® Sound Level Meter
Sound Meters
Sound level meter features three decibel ranges for greater precision: low (30–80dB), medium (50–100dB), and high (80–130dB).

  • Adjustable meter response time
  • Memory functions store readings for later display
  • Traceable®

Resolution is 0.1dB and accuracy is ±2dB

The response time on the meter may be set to fast (0.2 seconds) or slow (1.5 seconds). A memory data-hold function stores the highest reading achieved for later display. Meter uses a 1.3 cm (1/2") condenser microphone, and a recorder jack captures readings (AC and DC 10mV/dB).

The meter provides weightings for both A (frequencies for 500 to 10kHz) and C (frequencies from 30 to 10kHz) to meet IEC 651 Type-2 standards.

Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Ordering information: Meter is supplied with a 9V alkaline battery and carrying case.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty