Would you like to log in again?
Polycarbonate Forceps, Clear, Troemner
Supplier: Troemner
80864382
12985-010EA 44.3 USD InStock
12985-010
Polycarbonate Forceps, Clear, Troemner
Forceps
Forceps are designed for proper handling of weights.
  • Made of PVC
  • Economical
  • Used for handling milligram weights and larger weights up to 100 g
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty