Would you like to log in again?
VWR® Talon® Support Stand, Cast Iron, U-Shaped Base
Supplier: VWR
VWR® Talon®
80862633
12985-078EA 67.74 USD InStock
12985-078
VWR® Talon® Support Stand, Cast Iron, U-Shaped Base
Support Stands
Support stand base is constructed of cast iron with black enamel finish.
  • Cast iron construction with black enamel
  • U-shaped base
  • Built-in support rod holder
  • For use with VWR® Talon® lab frames
Base features three built-in support rod holders for additional support or to customize your stand. Locking knob accepts a 1.3 cm (1/2") diameter support rod (sold separately).
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty