Would you like to log in again?
SP Bel-Art Precision Plastic Dial Calipers, Bel-Art Products, a part of SP
Supplier: Bel-Art
H13416-0001
13196-070EA 173.53 USD InStock
13196-070
SP Bel-Art Precision Plastic Dial Calipers, Bel-Art Products, a part of SP
Calipers
Dial type calipers measure up to 150 mm in 0.1 mm increments.
  • Dial-type design
  • Non-conducting
  • Corrosion resistant
  • Lightweight for easy handling
  • Rigid plastic construction
A 33 mm (1.3") diameter dial is rotatable beneath the pointer allowing for ‘differential’ measurements. Black numbers on a white background afford effortless readability. These calipers are non-conducting, corrosion resistant, and lightweight for easy handling. Molded-in rigid plastic, they will not scratch like metal calipers can.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty