Would you like to log in again?
SP Bel-Art Round-Bottom Flask Support, Bel-Art Products, a part of SP
Supplier: Bel-Art
F38950-0000
13196-074CS 285.15 USD InStock
13196-074
SP Bel-Art Round-Bottom Flask Support, Bel-Art Products, a part of SP
Supports Flask Supports
Round-bottom flasks of any size up to 10 liters are cradled securely in the stepped concentric rings of this holder that can be stacked together for convenient storage.
  • Round-bottomed design
  • Polypropylene construction
  • Corrosion-resistant
  • Autoclavable
Polypropylene plastic rack is not affected by spilled acids, alkalis or other corrosive liquids. Steam autoclavable at 121°C (250°F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty