Would you like to log in again?
SP Bel-Art Magnetic Stirring Bar Arrester, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F37786-0000
13197-258EA 33.99 USD InStock
13197-258
SP Bel-Art Magnetic Stirring Bar Arrester, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
The Magnetic Stirring Bar Arrester holds your stirring bar in place when pouring from a beaker or flask by placing the arrester against the bottom of the container.
  • Polyethylene construction
The arrester is 9.5 mm (3/8") thick polyethylene, 51 mm (2") diameter with a radial magnet.
Ordering information: Other sizes can be made to order.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty