Would you like to log in again?
SP Bel-Art Poly-Hand Pumps, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
H32886-0000 H32787-0000 H32887-0000
13197-368EA 303.17 USD InStock
13197-368 54809-758 47742-770
SP Bel-Art Poly-Hand Pumps, Bel-Art Products, a part of SP
Pumps Drum/transfer Pumps
The Poly-Hand pump threads directly to a standard drum bung or carboy.
  • Polyethylene construction
  • Rugged and inert
  • Ergonomically designed handle for comfortable use
It is constructed entirely of rugged polyethylene and is inert to strong acids, alkalis and many solvents
The pump delivers by positive pressure on the downstroke of the piston and does not pressurize the vessel. Its ergonomic 'T' handle fits comfortably in a gloved hand. The pump will efficiently deliver against a 3.65 m (12 ft) head of water and empty a drum or carboy to within 3 mm (⅛") of the bottom.
Sizes are available to fit 114 L and 220 L (30/50 gal) plastic or steel drums and 20 L (5 gal.) carboy (16126-845 ).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty