Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Bel-Art Liquid Nitrogen Cooled Mortar, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
H37260-0001 H37260-0000
13197-414EA 721.64 USD InStock
13197-414 16126-652
SP Bel-Art Liquid Nitrogen Cooled Mortar, Bel-Art Products, a part of SP
Mortars and Pestles Mortars
Grind frozen samples without liquid nitrogen hazards.
  • Stainless steel mortar construction
  • Nested in heavily insulated polyethylene housing
  • Suction cup feet
  • Polyethylene handle included on pestle
A 304 stainless steel mortar is suspended by three brackets above a stainless steel bowl which holds liquid nitrogen
The housing measures 16.5 cm (6½”) in diameter x 11.4 cm (4½”) high and has three suction cup feet. The recommended size pestle is any size appropriate for a 275 mL mortar. Pestle not included.
The bowl is nested in a heavily insulated blue polyethylene housing. The mortar has an insulated handle, 10.2 cm (4”) long, that is removable for saving space.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty