Would you like to log in again?
SP Bel-Art Vac-Ring™ Filter Seal, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
H38959-0000
13197-444EA 39.9 USD InStock
13197-444
SP Bel-Art Vac-Ring™ Filter Seal, Bel-Art Products, a part of SP
Seals Sealing Rings
Slips easily onto Buchner funnel stems and seals-off the filter flask mouth to prevent air leaks during vacuum filtration.
  • Chemically resistant
  • Fits up to 22 mm diameter funnel stems
Made of black neoprene, the 9.5 mm thick by 76 mm O.D. ring is wide enough to fit filter flasks up to 4000 mL.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty