Would you like to log in again?
WETTASK™ Refillable Wiper Systems, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
WETTASK®
06471 06006 06471 06006 06211 77320 06211 06411 77320 06001 06411
89096-628CS 172.32 USD InStock
CA21903-014 89096-628 CA21903-012 CA89096-628 21903-014 21903-012 CA89194-380 CA89003-026 89194-380 89003-026 13197-506
WETTASK™ Refillable Wiper Systems, KIMBERLY-CLARK PROFESSIONAL®
Wipes Disposable Wipes and Towels Presaturated Wipes
The WETTASK™ Refillable wiper system is an easy-to-use and incredibly versatile alternative to using a spray bottle or open bucket in combination with a rag.
  • System portability enables users to spot clean germ 'hot zones' without the hassle of a full cleaning cart
  • Controlled chemical usage keeps costs low and elliminates waste
  • No laundering is required
  • Offers multiple wiping materials designed for the chemical solutions required to fit specific cleaning tasks
  • Solvents, Detergents, and Water-Based Cleaners utilize HYDROKNIT™ material
  • Compact, self-contained wet wiping system is safe, simple, and reliable
The enclosed system design helps to avoid contamination of the wipers and cleaning solution when in use. This reduces exposure to chemical vapors and splash, as well as the risk of chemical spills. Systems allow users the versatility of adding preferred disinfecting solutions, and preparing buckets ahead of time.
Three different wipers are available, depending upon the chemical solution being used. This hydro-entangled, cellulose/polypropylene material is manufactured without chemical binders or adhesives. Disinfectants and Sanitizers utilize spunlace material. Bleach, Disinfectants, and Sanitizers utilize meltblown material, which is compatible with Quaternary Amine Disinfectants.
Wipers provide a cloth-like feel. Typical system applications include surface preparation, surface cleaning, sanitizing, disinfecting, cleaning and removing soil, oil, or grease, and solvent application. Wiper color: white.
Ordering information: Cases are available with or without a bucket or cannister included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty