Would you like to log in again?
VWR® Modular Heating Sand Baths
Supplier: VWR
VWR®
80863137 80863134 80863135 80863133 80863136
13259-264EA 495.98 USD InStock
13259-264 13259-262 13259-270 13259-268 13259-266
VWR® Modular Heating Sand Baths
Block Heaters Blocks
For use in heating blocks, these sand baths are ideal for irregular vessels.
  • Now with Best-in-Class, Exclusive 5-Year Warranty
Stainless steel construction for superior corrosion resistance. Designed to hold sand, shot, or nonvolatile liquids. Right angle thermometers (61220-740 and 61220-742) have an accuracy rating of ± 1°C.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty