Would you like to log in again?
VWR® Modular Heating Block for Square Cuvettes
Supplier: VWR
VWR®
80863114
13259-290EA 223.23 USD InStock
13259-290
VWR® Modular Heating Block for Square Cuvettes
Block Heaters Blocks
Designed for 12 cuvettes, these blocks feature two parallel slots which fit six cuvettes in each slot, arranged side by side for excellent stability and heat transfer.
  • Now with Best-in-Class, Exclusive 5-Year Warranty
Slots accommodate 12.5mm (1/2") cuvettes. Single block size.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty