Would you like to log in again?
VWR® Bath Fluids
Supplier: VWR
VWR®
060326 060328 60330
13272-034EA 256.14 USD InStock
13272-034 13270-792 13270-794
VWR® Bath Fluids
Cleaning Agents Water Bath Protection Reagents
These heat-transfer fluids provide superior thermal stability for high or low temperatures.
  • Silicone heat transfer fluid
Check compatibility of fluids before using, as fluids are incompatible with Buna or natural rubber.
Packaging: Bottles are packaged in a carton for shipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty