Would you like to log in again?
VWR® Flow-Through and Immersion Coolers
Supplier: VWR
VWR®
F25N0V101B P10N6V101B P40N7V101B P35N5V101B P60N2V101B
89400-956EA 7186.86 USD InStock
89400-956 89400-960 13271-504 13271-502 13271-500
VWR® Flow-Through and Immersion Coolers
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Enhance and simplify your low-temperature laboratory work with these low temperature coolers.
  • Compact add-on cooling
  • VWR® Two-Year Limited Parts and Labor Warranty
Available as immersion probe and flow-through models, these units provide rapid, low-cost cooling of liquids to temperatures as low as -100°C
Flow-through Style Coolers offer an extremely economical alternative to tap water cooling. Use with heated circulating baths to reach temperatures below ambient. Use with refrigerated circulating baths to increase cooling capacity.
Immersion Probe Style Coolers are available with different fixed temperatures. Excellent for trapping and Dewar-type applications, these coolers are designed to run at maximum cooling and reduce the expense of using dry ice or liquid nitrogen. A flexible hose allows convenient placement of the cooling probe.
Typical applications include the cooling of exothermic reactions, freeze point determinations, freeze drying, impact testing, lyophilization and vapor and solvent trapping.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty