Would you like to log in again?
BOTTLE SS 5L 5.1CM TRICLAMP
Supplier: EAGLE STAINLESS CONTAINER
Stainless Steel Bottle^ Volume: 5L. Constructed of T316L stainless steel. With 5.1cm tri-clamp, standard 24 Ra interior/exterior finish, all welds seamless and polished, and with 18 gauge side/bottom wall thickness. For storage, shipping, or transfer of liquids and solid products.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BOTTLE SS 5L 5.1CM TRICLAMP 13308-172 PSF-18-316L-A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty