Would you like to log in again?
Capillary Tubes, Drummond
Supplier: Drummond
1-000-800/12
13450-216CS 141.36 USD InStock
13450-216
Capillary Tubes, Drummond
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Glass capillary tubes for coagulation, hemoglobin, prothrombin, blood grouping and typing.
  • Capacity 9.4 µL
  • Length 7.5cm
  • Internal Diameter 0.4-0.6mm.
Ordering information: Packaged in vials of 100 tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty