Would you like to log in again?
DuPont™ Tychem® 2000 Sleeves
Supplier: DuPont
Tychem®
QC500BYL00020000 QC500BYL0000
13450-338CS 339.26 USD InStock
CA13452-680 13450-338
DuPont™ Tychem® 2000 Sleeves
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Elastic at both ends
  • Bound seams
  • One size fits most
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing.
These sleeves feature elastic at both bicep, measuring 81/4", and wrist, measuring 61/2".
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty