Would you like to log in again?
Hazardous Chemical Pourer, Dynalon
Supplier: Dynalon
310374-0003 310374-0004 310374-0002
13450-604EA 148.49 USD InStock
13450-604 13450-602 13450-606
Hazardous Chemical Pourer, Dynalon
Funnels Dropping Funnels
The hazardous and corrosive chemical pourer is manufactured fully from PTFE and features completely inert pouring surfaces
Even the neck-sealing ring is encapsulated in PTFE. The unique design allows direct and controlled pouring due to the incorporated breather vent. The pourer comes in four sizes to fit standard reagent bottles with neck sizes of GL45, 38, 32, and 30. A captured PTFE closure ensures contaminant free contents when not in use. Bottle not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty