Would you like to log in again?
Graduated Bottles, Polypropylene, Dynalon
Supplier: Dynalon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
501505-0250 501505-0060 501505-0500 501505-0125 501505-1000
30620-170CS 474.98 USD InStock
30620-170 13450-622 13450-620 13450-626 13450-628
Graduated Bottles, Polypropylene, Dynalon
Bottles Plastic Bottles
High performance polypropylene bottles and closures are ideal for use with a wide range of liquids and solids.
  • Autoclavable leak-proof construction
  • High clarity polypropylene allows easy viewing of the fluid level
Full range of sizes with high chemical resistance, printed with large white marker space for clear, easy, and safe labeling. Wide mouth for easy access filling and clear crisp printed graduations show milliliters and fluid ounces, allowing estimation of volumes remaining and/or decanted.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty