Would you like to log in again?
DuPont™ Tychem® 4000 Coveralls with Laydown Collar and Elastic Wrists and Ankles, Bound Seams
Supplier: DuPont
Tychem®
SL125BWH4X001200 SL125BWHXL001200 SL125BWH3X001200 SL125BWH3X001200 SL125BWHLG001200 SL125BWHMD001200 SL125BWHMD00 SL125BWHLG00 SL125BWH2X00 SL125BWH6X001200 SL125BWH2X001200 SL125BWHXL00 SL125BWH4X00 SL125BWH5X001200
13451-066CS 395.77 USD InStock
CA13451-076 13451-066 CA13451-066 13451-076 CA13451-074 13451-068 CA13451-068 10016-484 10016-794 10016-486 13451-070 CA13451-072 13451-072 CA13451-070
DuPont™ Tychem® 4000 Coveralls with Laydown Collar and Elastic Wrists and Ankles, Bound Seams
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Laydown collar
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications include chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments. Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration. Coveralls feature a laydown collar, and elastic openings at wrists and ankles for a tighter fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty