Would you like to log in again?
DuPont™ ProClean® Boot Covers with PVC Sole
Supplier: DuPont
ProClean®
PC444SWHLG01000B PC444SWHLG0B PC444SWHXL0B PC444SWHXL01000B PC444SWHMD01000B PC444SWHMD0B
13451-210CS 235.01 USD InStock
13451-210 CA13451-212 13451-212 CA13451-210 CA13451-216 13451-216
DuPont™ ProClean® Boot Covers with PVC Sole
Disposable Garments
ProClean® offers improved non-hazardous light liquid splash protection.
  • Serged seams
  • PVC sole
  • Covered elastic opening
  • Elastic ankles
ProClean® is made from a microporous composite fabric. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic openings and elastic at ankles allow for a secure fit. PVC soles provide skid-resistance and durability.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty