Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Antistatic Weighing Canoes, Dyn-A-Med Products
Supplier: Dyn-A-Med Products
80056 80056 80051 80061
470202-542CS 408.65 USD InStock
470202-542 13451-990 13451-986 13451-989
Antistatic Weighing Canoes, Dyn-A-Med Products
Boats Weighing Boats
Anti-static Weighing Canoes are designed for accurate weighing of static-affected material.
  • Disposable
  • Polystyrene construction
  • Anti-static finish
Weighing Canoes are made of recyclable virgin polystyrene with 109 ohm2 resistivity. Contoured sides provide a convenient non-slip gripping surface. Canoe shape allows easy pouring of weighed samples. Available in three sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty