Would you like to log in again?
Dupont™ ProShield® 60 Coveralls with Laydown Collar and Open Wrists
Supplier: DuPont
Pro Shield
NG120SWH3X002500 NG120SWH2X00 NG120SWHMD00 NG120SWH5X002500 NG120SWH3X00 NG120SWHSM002500 NG120SWH6X002500 NG120SWH4X00 NG120SWH2X002500 NG120SWHXL00 NG120SWHLG00 NG120SWHSM00 NG120SWH6X00 NG120SWHLG002500 NG120SWHMD002500 NG120SWH5X002500 NG120SWH4X002500 NG120SWHXL002500
13452-258CS 188.54 USD InStock
13452-258 13452-256 13452-266 13452-254 13452-264 13452-252 13452-262 CA13452-260 13452-250 13452-260 CA13452-250 CA13452-262 CA13452-252 CA13452-264 CA13452-254 CA13452-266 CA13452-256 CA13452-258
Dupont™ ProShield® 60 Coveralls with Laydown Collar and Open Wrists
Disposable Garments
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Laydown collar
  • Open wrists and ankles
  • Serged seams
ProShield® 60 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation. Coveralls feature a laydown collar and open wrists and ankles.
Certifications: Thickness meets ASTM D751.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty