Would you like to log in again?
Dupont™ ProShield® 60 Coveralls with Standard Hood and Attached Skid Resistant Boots
Supplier: DuPont
Pro Shield
NG122SWHLG00 NG122SWH4X002500 NG122SWH4X00 NG122SWHSM002500 NG122SWH2X002500 NG122SWHSM002500 NG122SWH5X002500 NG122SWH6X002500 NG122SWHXL00 NG122SWH3X002500 NG122SWH3X00 NG122SWHMD00 NG122SWH5X00 NG122SWHMD002500 NG122SWHXL002500 NG122SWHLG002500 NG122SWH6X002500 NG122SWH2X00
10016-106CS 217.76 USD InStock
10016-106 CA10016-104 13452-288 13452-286 CA13452-290 13452-296 CA13452-292 13452-284 13452-294 CA13452-282 CA13452-294 13452-292 CA13452-284 CA13452-296 CA13452-286 13452-290 10016-104 CA13452-288
Dupont™ ProShield® 60 Coveralls with Standard Hood and Attached Skid Resistant Boots
Disposable Garments
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Standard fit hood
  • Elastic wrists
  • Attached skid-resistant boots
  • Serged seams
ProShield® 60 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation. Attached hood with elastic around face opening is standard fit. Front zipper closure allows for easy donning and doffing. Elastic opening at wrists allows for a tighter fit. Attached ProShield® 3 boots have a special coating to provide added skid-resistance.
Certifications: Thickness meets ASTM D751.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty