Would you like to log in again?
DuPont™ ProClean® Frocks, Front Zipper Closure
Supplier: DuPont
ProClean®
PC271SWH4X0B PC271SWH4X00300B PC271SWH2X00300B PC271SWHMD0B PC271SWHLG00300B PC271SWHLG0B PC271SWHMD00300B PC271SWH2X0B PC271SWHSM00300B PC271SWH3X0B PC271SWHXL00300B PC271SWHSM00300B PC271SWHXL0B PC271SWH3X00300B
13452-548CS 236.02 USD InStock
13452-548 13452-546 13452-544 13452-554 13452-552 13452-550 CA89129-704 89129-704 CA13452-550 CA13452-552 CA13452-554 CA13452-544 CA13452-546 CA13452-548
DuPont™ ProClean® Frocks, Front Zipper Closure
Disposable Garments
ProClean® offers improved non-hazardous light liquid splash protection.
  • Serged seams
  • Bound neck
  • Set sleeve design
  • Covered elastic wrists
  • Front zipper closure
ProClean® is made from a microporous composite fabric. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Covered elastic openings at wrists allows for a tighter fit. Bound neck ensures lower particle shedding. Frocks provide body-length jacket design with front zipper closure for easy donning and doffing.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty