Would you like to log in again?
DuPont™ Tychem® 2000 Coveralls with Laydown Collar and Open Wrists and Ankles, Serged Seams
Supplier: DuPont
Tychem®
QC120SYL4X001200 QC120SYL4X001200 QC120SYLLG001200 QC120SYLLG001200 QC120SYL2X001200 QC120SYL3X001200 QC120SYL6X001200 QC120SYLXL001200 QC120SYL3X001200 QC120SYL5X00 QC120SYLXL001200 QC120SYL7X001200 QC120SYLMD001200 QC120SYL2X001200 QC120SYL5X001200 QC120SYLMD001200
10010-186CS 193.02 USD InStock
CA10010-186 10010-186 CA10010-188 10010-184 CA10010-182 CA10010-184 10010-188 13452-630 10016-366 10016-368 CA13452-630 10010-190 CA10010-190 CA10010-192 10010-182 10010-192
DuPont™ Tychem® 2000 Coveralls with Laydown Collar and Open Wrists and Ankles, Serged Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Laydown collar
  • Open wrists and ankles
  • Serged seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls feature a laydown collar, open wrists, and open ankles.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty