Would you like to log in again?
DuPont™ Tychem® RESPONDER® CSM Coveralls with Attached Gloves and Socks
Supplier: DuPont
RC128TTNLG00 RC128TTNXL00 RC128TTNLG000100 RC128TTN3X000100 RC128TTNSM00 RC128TTN3X00 RC128TTN4X000100 RC128TTNMD000100 RC128TTNSM000100 RC128TTNXL000100 RC128TTN2X000100 RC128TTNMD00 RC128TTN2X00
13452-706CS 763.95 USD InStock
13452-706 CA13452-706 13452-704 13452-702 13452-700 13452-698 13452-696 76187-838 CA13452-696 CA13452-698 CA13452-700 CA13452-702 CA13452-704
DuPont™ Tychem® RESPONDER® CSM Coveralls with Attached Gloves and Socks
Disposable Garments
Tychem® RESPONDER® CSM combines multiple film barriers laminated to both sides of a high-strength polypropylene substrate for high level protection against toxic and corrosive chemicals.
  • Respirator fit hood
  • Replaceable attached butyl gloves
  • Attached socks with outer boot flaps
  • Double storm flap with hook & loop closure
  • Double taped seams
It is effective for handling chemical warfare agents as well as military site cleanup and HazMat first response. Tychem® RESPONDER® CSM fabric provides at least 30 minutes of protection against 320 chemical challenges.
Each suit undergoes rigorous inspection and each Level A suit, pressure testing. Each fabric lot is tested using live toxic agents plus one garment for each test fabric production run is also tested for seam barrier with live toxic agents. Coveralls are a desert tan color for low visibility applications.
Attached respirator fit hood with elastic around face opening is designed to cover the neck and the chin and to fit around a respirator face mask. Coveralls feature a strong zipper for tough, flexible closure that is longer to extend all the way to the chin for complete coverage of neck area.
Storm flap covers zipper, which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper. Attached MIL-G-12223 Butyl gloves provide mechanical and physical durability with vapor tight ring and clamp system and are replaceable. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Certifications: Thickness meets ASTM D1117. Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty