Would you like to log in again?
DuPont™ Tyvek® 400 Frocks with Laydown Collar and Elastic Wrists, Extra Long
Supplier: DuPont
Tyvek®
TY211SWH4X003000 TY211SWH3X003000 TY211SWHSM003000 TY211SWH5X00 TY211SWHXL003000 TY211SWHSM00 TY211SWHLG003000 TY211SWHLG00 TY211SWH6X003000 TY211SWH2X00 TY211SWH4X00 TY211SWHMD00 TY211SWHXL00 TY211SWH5X003000 TY211SWH7X003000 TY211SWH2X003000 TY211SWH3X00 TY211SWHMD003000
13452-894CS 400.88 USD InStock
13452-894 10016-924 10016-926 CA13450-510 13450-512 13450-510 CA13452-894 CA13450-516 13450-506 13450-516 CA13450-506 13450-504 CA13450-508 13450-514 CA13450-512 CA13450-514 13450-508 CA13450-504
DuPont™ Tyvek® 400 Frocks with Laydown Collar and Elastic Wrists, Extra Long
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Laydown collar
  • Elastic wrists
  • Extends to knee
  • Front snap closure
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Frocks feature body-length jacket design with front snap closures for easy donning and doffing. Ideal for applications that do not require the full body coverage of a coverall. Frocks also feature a laydown collar and elastic openings at wrists for a tighter fit.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty