Would you like to log in again?
DuPont™ ProShield® 6 SFR Coveralls with Standard Hood
Supplier: DuPont
ProShield®
TM127SBU3X002500 TM127SBU5X002500 TM127SBUXL00 TM127SBUXL002500 TM127SBU3X002500 TM127SBU2X002500 TM127SBUMD002500 TM127SBU4X002500 TM127SBU7X002500 TM127SBULG002500 TM127SBU4X002500 TM127SBUMD002500 TM127SBU2X002500 TM127SBU6X002500
13452-978CS 345.02 USD InStock
13452-978 13452-976 10016-568 13452-972 13452-982 13452-980 10016-572 CA13452-980 10016-566 CA13452-982 CA13452-972 CA13452-976 CA13452-978 10016-570
DuPont™ ProShield® 6 SFR Coveralls with Standard Hood
Disposable Garments
ProShield® 6 SFR is a cost-efficient disposable overgarment designed to help protect and preserve primary flame resistant garments.
  • Standard fit hood
  • Elastic wrists and ankles
  • Front zipper closure
  • Serged seams
ProShield® 6 SFR garments are intended to be worn over primary flame resistant garments to provide a barrier against non-hazardous particles and aerosols. ProShield® 6 SFR garments are designed to not contribute to additional burn injury in the event of an industrial fire or electric arc event. ProShield® 6 SFR garments will not ignite and continue to burn after the flame source is removed. ProShield® 6 SFR garments are an excellent choice as a cost efficient cover to protect reusable FR garments. Applications include Steel Mills, Coal Mines, Maintenance Work, Refineries, Laboratories, and Utilities. Coveralls feature an attached hood and elastic openings at wrists and ankles for a tighter fit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty