Short-Term detector tubes for use with the accuro® Gas Detector Pump and Aerotest Breathing Air Tester.
- Dräger Tubes are glass vials filled with a chemical reagent that reacts to a specific chemical or family of chemicals
- A variety of tubes are available for the measurement of numerous different gases and vapors
Ordering information: For additional tubes, contact your Avantor sales representative.