Would you like to log in again?
UNION BODY 1/2-20 1/8IN THRU
Supplier: IDEX HEALTH & SCIENCE LLC
Union Body^ For 1/4 inch OD tubing 1/2-20 threads, .188 inch (4.8mm) thru-hole, PEEK*, black, no fittings
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
UNION BODY 1/2-20 1/8IN THRU 13500-026 U-658-01 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty