Would you like to log in again?
Nalgene® Diagnostic Bottles, White HDPE, without Closures, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
362008-9125 362008-9025 362008-9050
16210-882CS 1049.28 USD InStock
16210-882 73521-322 13500-126
Nalgene® Diagnostic Bottles, White HDPE, without Closures, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene white HDPE diagnostic bottles without closures are used in test kits and many small-volume applications.
They can be used for sample storage and shipping of small volume, valuable reagents. Diagnostic bottles can be used for sample storage, and shipping of valuable reagents that are light sensitive.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty