Would you like to log in again?
Nalgene® Boston Bottles, Round, Opaque White, HDPE, without Closures, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
362008-0001 362008-0016 362008-0008 362008-0032 362008-0004 362008-0002
13500-130CS 446.86 USD InStock
13500-130 13500-132 16210-880 13500-134 13500-136 13500-128
Nalgene® Boston Bottles, Round, Opaque White, HDPE, without Closures, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene boston round opaque white HDPE bottles are specifically designed for demanding lab use, and have durable uniform walls that are resistant to splitting or puncturing.
They are reliable, durable, chemical resistant, and are opaque white for light sensitive products. They also meet the requirements of light-resistant containers per USP latest edition.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty