Would you like to log in again?
Four-Point Suspension Hard Hats, Honeywell Safety
Supplier: Honeywell Safety
A79R010000 A79R020000 A79R150000 A59010000 A79R070000 A59010000 A59030000 A59040000 A79R030000 A79R040000 A79010000 A59070000 A59040000 A59020000
13502-137CS 356.69 USD InStock
13502-137 13502-139 CA13502-200 CA13502-201 13502-133 13502-157 13502-200 CA13502-197 CA13502-195 13502-131 13502-142 CA13502-193 13502-141 13502-193
Four-Point Suspension Hard Hats, Honeywell Safety
Head Protection Safety Helmets
Peak hard hats feature a grooved high-density polyethylene outer shell, rain trough, and accessory slots
  • Grooved high-density polyethylene outer shell, rain trough, and accessory slots
  • Ratchet or pin lock adjustments
  • Suspension offers three-level height adjustment
Four-point hats come with ratchet or pin lock adjustments. Suspension offers a three-level height adjustment.
Certifications: CE marked. Meet ANSI Type I Class E requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty