Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Face Shield Visors, Honeywell Safety

Supplier: Honeywell Safety
A8154 A8150/40 A8154 A8150/40
13502-275CS 70.91 USD InStock
CA13502-275 CA63010-422 13502-275 13502-274
Face Shield Visors, Honeywell Safety
Face Shields
Fits headgear and brackets with universal slot design. Available as a flat, die cut visor made of PETG. Suitable for eye and face protection. Also available as a formed, preshaped visor made of clear polycarbonate, suitable for heat deflection up to 121 °C (250 °F) continuous.

  • Fits headgear and brackets with universal slots design
  • Suitable for heat deflection up to 121 °C (250 °F) continuous

Thickness: 1 cm (0.4")
Dimensions: 20.3 Lx39.2 W cm (8x15.5").

Certifications: Meets ANSI Z87.1-1989. CSA certified.

Ordering information: Visor orders do not include brackets or mounting material. Brackets and headgear must be purchased separately.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty