Would you like to log in again?
Plastic Bracket for Hard Hats, Honeywell Safety
Supplier: Honeywell Safety
CP5006 CP5005 CP5005 CP5006
13502-278EA 15.3 USD InStock
CA13502-278 13502-278 13502-281 13502-280
Plastic Bracket for Hard Hats, Honeywell Safety
Head Protection Safety Helmets
Lightweight plastic bracket fits into any slotted hard hat
Bracket 13502-278 fits Everest wide brim hats only (see 13502-144 series).
Bracket accommodates ORIS® earmuff systems. Bracket cam lock system accepts a wide range of face shields.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty