Would you like to log in again?
Gas Detector Tubes for Acetone, Sensidyne
Supplier: Sensidyne
102SA
13502-652PK 211.61 USD InStock
13502-652
Gas Detector Tubes for Acetone, Sensidyne
Gas Detection Systems
Gas detector tubes are used for detection of airbourne gases and vapors.
  • Contains an antibacterial grip
  • Easy to use
These detector tubes safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors at a very low cost-per sample. These tubes are easy to use and provide a low-cost method of routine or spot sampling for gases and vapors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty