Would you like to log in again?
Whatman™ UNIFILTER® Microplates, 96-Well, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
UNIFILTER® Whatman™ UNIFILTER®
7700-2810 7770-0062 7700-2804 7700-2808 7770-0062 7700-2801 7700-2805 7700-2805 7700-2803 7700-2804 7700-2801 7700-2803 7700-2808 7700-2810
13503-364PK 1273.31 USD InStock
CA13503-038 CA13503-364 CA13503-034 13503-364 13503-034 CA13503-048 CA13503-052 CA13503-042 13503-038 13503-048 CA13503-040 13503-042 13503-052 13503-040
Whatman™ UNIFILTER® Microplates, 96-Well, Whatman products (Cytiva)
Microplates Filter Plates
UNIFILTER 96-well microplate, 800 µl, long drip director.
  • No cross talk: Patented integral filter design prevents well-to-well cross contamination
  • Economical to use: Wide range of well volume options ensures efficient use of materials
  • Better control: Choice of filter media allows control of the flow rates and retention characteristics
  • Versatile: A broad range of filtration media is available, including glass fiber, polypropylene, cellulose nitrate, cellulose acetate, nylon, and ion exchange cellulose
The 800 µl UNIFILTER microplate is most typically used in purifications, isolations, and separations of biomolecules, particularly DNA.
The microplate has a well volume of 800 μl, which is excellent for standard DNA plasmid miniprep chemistries. The choice of short or long drip directors is application-specific. UNIFILTER 800 μl is constructed from rigid high-grade polystyrene.
Proprietary Whatman UNIFILTER microplates with filter bottom wells are convenient and ready to use. Available in 24-, 96-, and 384-well formats, UNIFILTER microplates offer a choice of filter media to meet exact application requirements.
The novel drip director design of Whatman UNIFILTER microplates ensures precise collection of the filtrate to allow for further processing and analysis.
Whatman UNIFILTER microplates from Cytiva's business are available in other plate materials and well formats. Well volumes range from 100 μl to 10 ml.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty